Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGATTCAATCACCCACCTACCCT[C/T]TAAAGTGGTGATACAAGATATTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616017 | ||||||||||||||||||||
Literature Links: |
TRIM69 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM69 - tripartite motif containing 69 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301144.1 | 426 | Intron | NP_001288073.1 | |||
NM_001301145.1 | 426 | Intron | NP_001288074.1 | |||
NM_001301146.1 | 426 | Intron | NP_001288075.1 | |||
NM_080745.4 | 426 | Intron | NP_542783.2 | |||
NM_182985.4 | 426 | Missense Mutation | TCT,TTT | S,F 28 | NP_892030.3 |