Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGTGTTTTCAGGCCACACTGAAC[A/G]GAATCTGAAATAAAGGAAAAGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607089 MIM: 177070 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNDBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCNDBP1 - cyclin D1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012142.4 | 1950 | Intron | NP_036274.3 | |||
XM_006720448.1 | 1950 | Intron | XP_006720511.1 |
EPB42 - erythrocyte membrane protein band 4.2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000119.2 | 1950 | Missense Mutation | CGT,TGT | R,C 670 | NP_000110.2 | |
NM_001114134.1 | 1950 | Missense Mutation | CGT,TGT | R,C 640 | NP_001107606.1 | |
XM_005254225.1 | 1950 | Missense Mutation | CGT,TGT | R,C 605 | XP_005254282.1 | |
XM_011521349.2 | 1950 | Missense Mutation | CGT,TGT | R,C 670 | XP_011519651.1 | |
XM_011521350.2 | 1950 | Missense Mutation | CGT,TGT | R,C 670 | XP_011519652.1 | |
XM_011521351.2 | 1950 | Missense Mutation | CGT,TGT | R,C 670 | XP_011519653.1 | |
XM_011521352.2 | 1950 | Missense Mutation | CGT,TGT | R,C 658 | XP_011519654.1 | |
XM_011521353.2 | 1950 | Missense Mutation | CGT,TGT | R,C 635 | XP_011519655.1 | |
XM_011521354.2 | 1950 | Missense Mutation | CGT,TGT | R,C 485 | XP_011519656.1 |