Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCGGGCTGGTGAGGAATGGAGC[A/C]GGTAGGCTGCTGCGGCGAGTGCCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609096 MIM: 612501 | ||||||||||||||||||||
Literature Links: |
FBXO22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO22 - F-box protein 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012170.3 | 33 | Missense Mutation | CAG,CCG | Q,P 3 | NP_036302.1 | |
NM_147188.2 | 33 | Missense Mutation | CAG,CCG | Q,P 3 | NP_671717.1 | |
XM_011521447.2 | 33 | UTR 5 | XP_011519749.1 |
UBE2Q2 - ubiquitin conjugating enzyme E2 Q2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |