Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACGGCCCGGCGCGTGGCTCGGG[C/T]TCGGGCGGAGCTGGAGACGTGTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DTWD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DTWD1 - DTW domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144955.1 | 18 | UTR 5 | NP_001138427.1 | |||
NM_020234.5 | 18 | UTR 5 | NP_064619.2 | |||
XM_011521815.1 | 18 | UTR 5 | XP_011520117.1 | |||
XM_017022419.1 | 18 | UTR 5 | XP_016877908.1 | |||
XM_017022420.1 | 18 | Intron | XP_016877909.1 | |||
XM_017022421.1 | 18 | Intron | XP_016877910.1 | |||
XM_017022422.1 | 18 | Intron | XP_016877911.1 | |||
XM_017022423.1 | 18 | UTR 5 | XP_016877912.1 | |||
XM_017022424.1 | 18 | UTR 5 | XP_016877913.1 | |||
XM_017022425.1 | 18 | UTR 5 | XP_016877914.1 | |||
XM_017022426.1 | 18 | Intron | XP_016877915.1 | |||
XM_017022427.1 | 18 | UTR 5 | XP_016877916.1 | |||
XM_017022428.1 | 18 | Intron | XP_016877917.1 | |||
XM_017022429.1 | 18 | Intron | XP_016877918.1 | |||
XM_017022430.1 | 18 | UTR 5 | XP_016877919.1 | |||
XM_017022431.1 | 18 | Intron | XP_016877920.1 |