Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGGGCACCATGGCGTCCATCCT[A/G]GATGAGTACGAGAACTCGCTGTCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608551 | ||||||||||||||||||||
Literature Links: |
VPS18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VPS18 - VPS18, CORVET/HOPS core subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020857.2 | 374 | Silent Mutation | CTA,CTG | L,L 5 | NP_065908.1 | |
XM_011521843.2 | 374 | Intron | XP_011520145.1 | |||
XM_011521844.2 | 374 | UTR 5 | XP_011520146.1 | |||
XM_017022447.1 | 374 | UTR 5 | XP_016877936.1 |