Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCAGGTAAACTTTTCGAACACAG[C/T]CTTGATTGGCACTGTCTTTTTTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608879 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS13C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VPS13C - vacuolar protein sorting 13 homolog C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018088.2 | 11208 | Intron | NP_001018098.1 | |||
NM_017684.4 | 11208 | Missense Mutation | GAC,GGC | D,G 3663 | NP_060154.3 | |
NM_018080.3 | 11208 | Intron | NP_060550.2 | |||
NM_020821.2 | 11208 | Missense Mutation | GAC,GGC | D,G 3706 | NP_065872.1 | |
XM_011521713.2 | 11208 | Intron | XP_011520015.1 | |||
XM_011521714.2 | 11208 | Intron | XP_011520016.1 |