Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTACCCCGAAGCGCTGCTACGATT[C/G]CCAGGGGGAGCGCCTCTCACACTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SLTM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLTM - SAFB like transcription modulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013843.2 | 2664 | Silent Mutation | GGC,GGG | G,G 901 | NP_001013865.1 | |
NM_024755.3 | 2664 | Silent Mutation | GGC,GGG | G,G 919 | NP_079031.2 | |
XM_006720686.3 | 2664 | Silent Mutation | GGC,GGG | G,G 972 | XP_006720749.3 | |
XM_006720690.1 | 2664 | Silent Mutation | GGC,GGG | G,G 488 | XP_006720753.1 | |
XM_011522022.1 | 2664 | Silent Mutation | GGC,GGG | G,G 1061 | XP_011520324.1 | |
XM_011522023.1 | 2664 | Silent Mutation | GGC,GGG | G,G 1043 | XP_011520325.1 | |
XM_011522024.1 | 2664 | Silent Mutation | GGC,GGG | G,G 1011 | XP_011520326.1 | |
XM_011522025.1 | 2664 | Silent Mutation | GGC,GGG | G,G 995 | XP_011520327.1 | |
XM_011522026.2 | 2664 | Silent Mutation | GGC,GGG | G,G 988 | XP_011520328.2 | |
XM_011522027.1 | 2664 | Silent Mutation | GGC,GGG | G,G 979 | XP_011520329.1 | |
XM_011522028.1 | 2664 | Silent Mutation | GGC,GGG | G,G 961 | XP_011520330.1 | |
XM_011522029.2 | 2664 | Silent Mutation | GGC,GGG | G,G 873 | XP_011520331.1 | |
XM_011522030.2 | 2664 | Intron | XP_011520332.1 | |||
XM_011522031.2 | 2664 | Silent Mutation | GGC,GGG | G,G 691 | XP_011520333.1 | |
XM_017022576.1 | 2664 | Silent Mutation | GGC,GGG | G,G 851 | XP_016878065.1 | |
XM_017022577.1 | 2664 | Silent Mutation | GGC,GGG | G,G 970 | XP_016878066.1 | |
XM_017022578.1 | 2664 | Silent Mutation | GGC,GGG | G,G 954 | XP_016878067.1 | |
XM_017022579.1 | 2664 | Silent Mutation | GGC,GGG | G,G 866 | XP_016878068.1 | |
XM_017022580.1 | 2664 | Silent Mutation | GGC,GGG | G,G 488 | XP_016878069.1 |