Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGGTAACCGCTGAGGTCGGTAT[A/G]GAAGAATGTGACCAGGTTGGTCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603035 MIM: 607050 | ||||||||||||||||||||
Literature Links: |
IL16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL16 - interleukin 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172128.1 | 1128 | Intron | NP_001165599.1 | |||
NM_004513.5 | 1128 | Intron | NP_004504.3 | |||
NM_172217.3 | 1128 | Intron | NP_757366.2 | |||
XM_005254342.3 | 1128 | Intron | XP_005254399.1 | |||
XM_005254346.4 | 1128 | Intron | XP_005254403.1 | |||
XM_011521519.2 | 1128 | Intron | XP_011519821.1 | |||
XM_011521520.2 | 1128 | Intron | XP_011519822.1 | |||
XM_017022140.1 | 1128 | Intron | XP_016877629.1 | |||
XM_017022141.1 | 1128 | Intron | XP_016877630.1 | |||
XM_017022142.1 | 1128 | Intron | XP_016877631.1 | |||
XM_017022143.1 | 1128 | Intron | XP_016877632.1 | |||
XM_017022144.1 | 1128 | Intron | XP_016877633.1 | |||
XM_017022145.1 | 1128 | Intron | XP_016877634.1 | |||
XM_017022146.1 | 1128 | Intron | XP_016877635.1 |
STARD5 - StAR related lipid transfer domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181900.2 | 1128 | Missense Mutation | CAT,TAT | H,Y 177 | NP_871629.1 | |
XM_006720703.1 | 1128 | Intron | XP_006720766.1 | |||
XM_017022639.1 | 1128 | Missense Mutation | CAT,TAT | H,Y 156 | XP_016878128.1 |