Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGGCATGAGGAATGGCCCCGG[A/C]TGGGATGTCAGGGGCCCAGGACAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613948 | ||||||||||||||||||||
Literature Links: |
SPATA8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPATA8 - spermatogenesis associated 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304804.1 | 260 | Intron | NP_001291733.1 | |||
NM_001304805.1 | 260 | Intron | NP_001291734.1 | |||
NM_001304806.1 | 260 | Intron | NP_001291735.1 | |||
NM_173499.4 | 260 | Missense Mutation | GAT,GCT | D,A 4 | NP_775770.1 | |
XM_011521265.2 | 260 | UTR 5 | XP_011519567.1 |
SPATA8-AS1 - SPATA8 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |