Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGAGTGCTGGCAAGGGCGGCCC[A/C]CAGGAGGAGGTGACCAGGGGACCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZSCAN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZSCAN2 - zinc finger and SCAN domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007072.1 | 421 | Silent Mutation | CCA,CCC | P,P 65 | NP_001007073.1 | |
NM_017894.5 | 421 | Silent Mutation | CCA,CCC | P,P 65 | NP_060364.4 | |
NM_181877.3 | 421 | Silent Mutation | CCA,CCC | P,P 65 | NP_870992.2 | |
XM_017022392.1 | 421 | Silent Mutation | CCA,CCC | P,P 65 | XP_016877881.1 | |
XM_017022393.1 | 421 | Silent Mutation | CCA,CCC | P,P 65 | XP_016877882.1 |