Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGTTGCTGCTGCGGGGTAAGCG[A/G]GAAAGACACCACACATTGCGCAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTDSPL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTDSPL2 - CTD small phosphatase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016396.2 | 102 | UTR 5 | NP_057480.2 | |||
XM_005254441.2 | 102 | Intron | XP_005254498.1 | |||
XM_006720565.2 | 102 | Intron | XP_006720628.1 | |||
XM_011521665.2 | 102 | UTR 5 | XP_011519967.1 | |||
XM_017022306.1 | 102 | Intron | XP_016877795.1 | |||
XM_017022307.1 | 102 | UTR 5 | XP_016877796.1 |