Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCTAGGACTCCACTAGCAGACT[A/G]TTCCTTGCCCTGCCCTCTCCTAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616844 MIM: 602437 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C15orf62 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C15orf62 - chromosome 15 open reading frame 62 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DNAJC17 - DnaJ heat shock protein family (Hsp40) member C17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GCHFR - GTP cyclohydrolase I feedback regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005258.2 | Intron | NP_005249.1 |