Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATGAAATATATTTAGTCTCTGT[C/T]CCTGGTACTTGTCACCTGGTTTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611695 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTBK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTBK2 - tau tubulin kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173500.3 | Intron | NP_775771.3 | ||||
XM_005254171.4 | Intron | XP_005254228.1 | ||||
XM_005254173.4 | Intron | XP_005254230.1 | ||||
XM_006720402.3 | Intron | XP_006720465.1 | ||||
XM_006720403.3 | Intron | XP_006720466.1 | ||||
XM_017021950.1 | Intron | XP_016877439.1 |