Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGCCTGTGTTGATAACTTAATC[C/T]GCAAGTCAAGTCTCTGCAGGAAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611796 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYSMD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYSMD2 - LysM domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143917.1 | 450 | Missense Mutation | CAG,CGG | Q,R 95 | NP_001137389.1 | |
NM_153374.2 | 450 | Missense Mutation | CAG,CGG | Q,R 186 | NP_699205.1 | |
XM_017022057.1 | 450 | Missense Mutation | CAG,CGG | Q,R 95 | XP_016877546.1 |
SCG3 - secretogranin III | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |