Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCTTCCCCTCTTCCCTTCAGAA[C/T]GAGACAGATGAGAGCCAGATCCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609585 MIM: 609548 MIM: 613472 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPLX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPLX3 - complexin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030005.2 | 349 | Silent Mutation | AAC,AAT | N,N 85 | NP_001025176.1 |
LMAN1L - lectin, mannose binding 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ULK3 - unc-51 like kinase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |