Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTACAAGCTCACACCCACCCATG[C/G]TGCCTCACCCGTCTACCGGCGCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612980 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IMP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IMP3 - IMP3, U3 small nucleolar ribonucleoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107984799 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX33 - sorting nexin 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318146.1 | 872 | Missense Mutation | GCT,GGT | A,G 259 | NP_001305075.1 | |
NM_153271.1 | 872 | Missense Mutation | GCT,GGT | A,G 259 | NP_695003.1 |