Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGAGGTACCAGGTCAGTGGCATGA[A/G]TGCTCTGCTCAGTTGCCTGTATAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603866 MIM: 170290 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEX11A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PEX11A - peroxisomal biogenesis factor 11 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271572.1 | 260 | Intron | NP_001258501.1 | |||
NM_001271573.1 | 260 | UTR 5 | NP_001258502.1 | |||
NM_003847.2 | 260 | Missense Mutation | ACT,ATT | T,I 75 | NP_003838.1 |
PLIN1 - perilipin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR93 - WD repeat domain 93 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |