Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGAACCAGGACAATGGCATTGA[C/T]GAGGGCTGGGTGAGCAGGCACCAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609748 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBL7 - ubiquitin like 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286739.1 | 666 | Missense Mutation | ATC,GTC | I,V 186 | NP_001273668.1 | |
NM_001286740.1 | 666 | Missense Mutation | ATC,GTC | I,V 186 | NP_001273669.1 | |
NM_001286741.1 | 666 | Missense Mutation | ATC,GTC | I,V 226 | NP_001273670.1 | |
NM_001286742.1 | 666 | Missense Mutation | ATC,GTC | I,V 226 | NP_001273671.1 | |
NM_032907.4 | 666 | Missense Mutation | ATC,GTC | I,V 186 | NP_116296.1 | |
NM_201265.1 | 666 | Missense Mutation | ATC,GTC | I,V 186 | NP_957717.1 | |
XM_011522124.2 | 666 | Missense Mutation | ATC,GTC | I,V 226 | XP_011520426.1 |