Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCGTACTTGAGCTTCTCCTCCAG[C/T]ACCTGCTTGCAGGAATACCTGGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605837 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HERC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HERC2 - HECT and RLD domain containing E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004667.5 | 7766 | Silent Mutation | GTA,GTG | V,V 4776 | NP_004658.3 | |
XM_005268276.4 | 7766 | Silent Mutation | GTA,GTG | V,V 4738 | XP_005268333.1 | |
XM_006720726.3 | 7766 | Silent Mutation | GTA,GTG | V,V 4771 | XP_006720789.1 | |
XM_006720727.3 | 7766 | Silent Mutation | GTA,GTG | V,V 4690 | XP_006720790.1 | |
XM_011522131.2 | 7766 | Silent Mutation | GTA,GTG | V,V 4615 | XP_011520433.1 | |
XM_011522133.2 | 7766 | Silent Mutation | GTA,GTG | V,V 3691 | XP_011520435.1 | |
XM_017022695.1 | 7766 | Silent Mutation | GTA,GTG | V,V 4738 | XP_016878184.1 | |
XM_017022696.1 | 7766 | Silent Mutation | GTA,GTG | V,V 4738 | XP_016878185.1 | |
XM_017022697.1 | 7766 | Silent Mutation | GTA,GTG | V,V 2498 | XP_016878186.1 | |
XM_017022698.1 | 7766 | Silent Mutation | GTA,GTG | V,V 2498 | XP_016878187.1 |