Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCGCCTTTGCTGCAACTGCCACT[C/T]TTCTAGTTCACGTTGCTGGAAATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610766 | ||||||||||||||||||||
Literature Links: |
MNS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MNS1 - meiosis specific nuclear structural 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018365.2 | 1450 | Missense Mutation | AAG,GAG | K,E 429 | NP_060835.1 |
TEX9 - testis expressed 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286449.1 | 1450 | Intron | NP_001273378.1 | |||
NM_198524.2 | 1450 | Intron | NP_940926.1 | |||
XM_005254359.4 | 1450 | Intron | XP_005254416.1 | |||
XM_005254361.3 | 1450 | Intron | XP_005254418.1 | |||
XM_011521530.2 | 1450 | Intron | XP_011519832.1 | |||
XM_017022160.1 | 1450 | Intron | XP_016877649.1 | |||
XM_017022161.1 | 1450 | Intron | XP_016877650.1 | |||
XM_017022162.1 | 1450 | Intron | XP_016877651.1 | |||
XM_017022163.1 | 1450 | Intron | XP_016877652.1 | |||
XM_017022164.1 | 1450 | Intron | XP_016877653.1 | |||
XM_017022166.1 | 1450 | Intron | XP_016877655.1 | |||
XM_017022167.1 | 1450 | Intron | XP_016877656.1 |