Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCCGGCCAACGTACCGTTCGGA[A/G]CTGTAGATGGAGATGCAGCGGCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607961 | ||||||||||||||||||||
Literature Links: |
MIR6881 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6881 - microRNA 6881 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA7A - semaphorin 7A (John Milton Hagen blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146029.2 | 1506 | Silent Mutation | AGC,AGT | S,S 509 | NP_001139501.1 | |
NM_001146030.2 | 1506 | Silent Mutation | AGC,AGT | S,S 358 | NP_001139502.1 | |
NM_003612.4 | 1506 | Silent Mutation | AGC,AGT | S,S 523 | NP_003603.1 |