Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGACTTGCCGTACTTTCTCTTGA[A/G]TTCAGACCTAATTTTCAACATGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 151740 | ||||||||||||||||||||
Literature Links: |
ANXA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANXA2 - annexin A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002857.1 | 1046 | Intron | NP_001002857.1 | |||
NM_001002858.2 | 1046 | Missense Mutation | CTC,TTC | L,F 325 | NP_001002858.1 | |
NM_001136015.2 | 1046 | Intron | NP_001129487.1 | |||
NM_004039.2 | 1046 | Intron | NP_004030.1 | |||
XM_011521477.2 | 1046 | Intron | XP_011519779.1 | |||
XM_017022090.1 | 1046 | Intron | XP_016877579.1 | |||
XM_017022091.1 | 1046 | Intron | XP_016877580.1 | |||
XM_017022092.1 | 1046 | Intron | XP_016877581.1 | |||
XM_017022093.1 | 1046 | Intron | XP_016877582.1 |