Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACAGGCCTTCTCTACATTCTTGG[C/T]CATTCCTGGAGGGCCGCAGCTGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606759 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUOX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUOX2 - dual oxidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014080.4 | 4770 | Missense Mutation | ACC,GCC | T,A 1523 | NP_054799.4 | |
XM_005254421.2 | 4770 | Missense Mutation | ACC,GCC | T,A 1523 | XP_005254478.1 |