Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCACACAGGCTCGAGCGTTTCCA[A/G]GTCAAGGGTAACGGAGCGTGGACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611254 MIM: 613298 | ||||||||||||||||||||
Literature Links: |
KIF7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIF7 - kinesin family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198525.2 | 3905 | Silent Mutation | ACC,ACT | T,T 1276 | NP_940927.2 | |
XM_011521531.2 | 3905 | Silent Mutation | ACC,ACT | T,T 1317 | XP_011519833.1 |
TICRR - TOPBP1 interacting checkpoint and replication regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308025.1 | 3905 | Intron | NP_001294954.1 | |||
NM_152259.3 | 3905 | Intron | NP_689472.3 |