Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGATGAAGTGGATGGCTTTGAAC[C/T]GGGCTGGGAAGGAATCCTGCGGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612196 MIM: 180090 | ||||||||||||||||||||
Literature Links: |
ABHD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABHD2 - abhydrolase domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RLBP1 - retinaldehyde binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000326.4 | 920 | Missense Mutation | NP_000317.1 | |||
XM_011521870.2 | 920 | Missense Mutation | XP_011520172.1 | |||
XM_017022460.1 | 920 | Missense Mutation | XP_016877949.1 |