Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTAATTTCCTAGAGATTCTGTTA[C/T]TTTTATTTTCAGTACTAAGCCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147582 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IREB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IREB2 - iron responsive element binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320941.1 | Intron | NP_001307870.1 | ||||
NM_001320942.1 | Intron | NP_001307871.1 | ||||
NM_001320943.1 | Intron | NP_001307872.1 | ||||
NM_004136.3 | Intron | NP_004127.1 |