Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACATTTTGCCCAAGTTCTAAGG[A/T]AAGTTCTGAAACTTAGTGGTGGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615940 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HACD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HACD3 - 3-hydroxyacyl-CoA dehydratase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016395.2 | 2287 | Intron | NP_057479.2 | |||
XM_011521664.1 | 2287 | Intron | XP_011519966.1 |
VWA9 - von Willebrand factor A domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136043.2 | 2287 | UTR 3 | NP_001129515.1 | |||
NM_001207058.1 | 2287 | UTR 3 | NP_001193987.1 | |||
NM_001207059.1 | 2287 | UTR 3 | NP_001193988.1 | |||
XM_006720708.3 | 2287 | UTR 3 | XP_006720771.1 | |||
XM_006720709.3 | 2287 | UTR 3 | XP_006720772.1 | |||
XM_006720711.2 | 2287 | UTR 3 | XP_006720774.1 | |||
XM_011522101.2 | 2287 | UTR 3 | XP_011520403.1 | |||
XM_017022644.1 | 2287 | UTR 3 | XP_016878133.1 |