Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTCCAGGTCCTGACGAGGTGAAC[C/G]GGTGTACTCTGAGAAGACGTAGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607249 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CATSPER2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CATSPER2 - cation channel sperm associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282309.1 | 1017 | Missense Mutation | NP_001269238.1 | |||
NM_001282310.1 | 1017 | Missense Mutation | CGT,GGT | R,G 274 | NP_001269239.1 | |
NM_054020.3 | 1017 | Missense Mutation | CGT,GGT | R,G 268 | NP_473361.1 | |
NM_172095.2 | 1017 | Missense Mutation | CGT,GGT | R,G 268 | NP_742093.1 | |
XM_017021904.1 | 1017 | Missense Mutation | CGT,GGT | R,G 268 | XP_016877393.1 |