Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACTGGATCTGGCAGATGGTGGC[C/T]GCGCTGAAGTCTCAGCCGGCCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151670 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIPC - lipase C, hepatic type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000236.2 | 317 | Silent Mutation | GCC,GCT | A,A 105 | NP_000227.2 | |
XM_005254372.1 | 317 | Silent Mutation | GCC,GCT | A,A 105 | XP_005254429.1 | |
XM_005254374.4 | 317 | Silent Mutation | GCC,GCT | A,A 117 | XP_005254431.2 | |
XM_006720502.3 | 317 | Silent Mutation | GCC,GCT | A,A 58 | XP_006720565.1 | |
XM_017022176.1 | 317 | Silent Mutation | GCC,GCT | A,A 117 | XP_016877665.1 |