Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGAGCACCGTGCTGACCTCCAA[A/C]TACCGTTAAGCTGGAGCCTCGGTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 141800 MIM: 141850 MIM: 609639 MIM: 142240 | ||||||||||||||||||||
Literature Links: |
HBA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HBA1 - hemoglobin subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HBA2 - hemoglobin subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000517.4 | 486 | Missense Mutation | AAA,AAC | K,N 140 | NP_000508.1 |
HBM - hemoglobin subunit mu | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HBQ1 - hemoglobin subunit theta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |