Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGGTGGTTTTTATATTCCTTGT[G/T]TGAGGGGTGTCTGTGATATTTGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160745 MIM: 609449 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYH11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYH11 - myosin heavy chain 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040113.1 | 6208 | UTR 3 | NP_001035202.1 | |||
NM_001040114.1 | 6208 | UTR 3 | NP_001035203.1 | |||
NM_002474.2 | 6208 | UTR 3 | NP_002465.1 | |||
NM_022844.2 | 6208 | UTR 3 | NP_074035.1 | |||
XM_011522502.2 | 6208 | Intron | XP_011520804.1 | |||
XM_017023250.1 | 6208 | Intron | XP_016878739.1 |
NDE1 - nudE neurodevelopment protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143979.1 | 6208 | Intron | NP_001137451.1 | |||
NM_017668.2 | 6208 | Intron | NP_060138.1 | |||
XM_005255396.4 | 6208 | Intron | XP_005255453.1 | |||
XM_006720897.3 | 6208 | Intron | XP_006720960.1 | |||
XM_006720900.3 | 6208 | Intron | XP_006720963.1 | |||
XM_011522553.2 | 6208 | Intron | XP_011520855.1 | |||
XM_017023349.1 | 6208 | Intron | XP_016878838.1 | |||
XM_017023350.1 | 6208 | Intron | XP_016878839.1 | |||
XM_017023351.1 | 6208 | Intron | XP_016878840.1 | |||
XM_017023352.1 | 6208 | Intron | XP_016878841.1 | |||
XM_017023353.1 | 6208 | Intron | XP_016878842.1 | |||
XM_017023354.1 | 6208 | Intron | XP_016878843.1 | |||
XM_017023355.1 | 6208 | Intron | XP_016878844.1 | |||
XM_017023356.1 | 6208 | Intron | XP_016878845.1 | |||
XM_017023357.1 | 6208 | Intron | XP_016878846.1 |