Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAAACAAACTGCTTGGTGAATT[G/T]TCACGAAGAGCAGATGGTGTGGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609463 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MKL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MKL2 - MKL1/myocardin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308142.1 | 115 | Intron | NP_001295071.1 | |||
NM_014048.4 | 115 | Intron | NP_054767.3 | |||
XM_005255452.3 | 115 | Intron | XP_005255509.1 | |||
XM_005255453.4 | 115 | UTR 5 | XP_005255510.1 | |||
XM_005255455.3 | 115 | Intron | XP_005255512.1 | |||
XM_006720908.3 | 115 | UTR 5 | XP_006720971.1 | |||
XM_006720909.3 | 115 | Intron | XP_006720972.1 | |||
XM_006720912.3 | 115 | Intron | XP_006720975.1 | |||
XM_006720913.2 | 115 | Intron | XP_006720976.1 | |||
XM_006720914.2 | 115 | Intron | XP_006720977.1 | |||
XM_011522568.2 | 115 | Intron | XP_011520870.1 | |||
XM_011522569.2 | 115 | Intron | XP_011520871.1 | |||
XM_017023501.1 | 115 | Intron | XP_016878990.1 | |||
XM_017023502.1 | 115 | Intron | XP_016878991.1 | |||
XM_017023503.1 | 115 | Intron | XP_016878992.1 | |||
XM_017023504.1 | 115 | Intron | XP_016878993.1 | |||
XM_017023505.1 | 115 | Intron | XP_016878994.1 |