Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACAGTGTGGCGAGCTACGAGAA[C/T]GAGGGTGCGTCTGGGATCCGAGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602354 MIM: 612583 | ||||||||||||||||||||
Literature Links: |
LAT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LAT - linker for activation of T-cells | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014987.1 | 688 | Silent Mutation | AAC,AAT | N,N 112 | NP_001014987.1 | |
NM_001014988.1 | 688 | Silent Mutation | AAC,AAT | N,N 111 | NP_001014988.1 | |
NM_001014989.1 | 688 | Silent Mutation | AAC,AAT | N,N 148 | NP_001014989.2 | |
NM_014387.3 | 688 | Silent Mutation | AAC,AAT | N,N 112 | NP_055202.1 |
SPNS1 - sphingolipid transporter 1 (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |