Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCTTTGCATGGGCGGCTTGTGCA[C/T]TCCCCTCACAATGTGAAGCACATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614031 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GFOD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GFOD2 - glucose-fructose oxidoreductase domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RANBP10 - RAN binding protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320238.1 | 3029 | UTR 3 | NP_001307167.1 | |||
NM_001320239.1 | 3029 | UTR 3 | NP_001307168.1 | |||
NM_001320240.1 | 3029 | UTR 3 | NP_001307169.1 | |||
NM_020850.2 | 3029 | UTR 3 | NP_065901.1 | |||
XM_005256070.2 | 3029 | UTR 3 | XP_005256127.1 | |||
XM_005256072.2 | 3029 | Intron | XP_005256129.1 | |||
XM_006721238.3 | 3029 | Intron | XP_006721301.1 | |||
XM_011523245.2 | 3029 | Intron | XP_011521547.1 | |||
XM_011523247.2 | 3029 | Intron | XP_011521549.1 | |||
XM_017023507.1 | 3029 | UTR 3 | XP_016878996.1 | |||
XM_017023508.1 | 3029 | Intron | XP_016878997.1 | |||
XM_017023509.1 | 3029 | UTR 3 | XP_016878998.1 |