Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTGCTTCTGTCACCGGCAGATT[C/G]GTCACCTGCACAGAGAGGAAATGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604470 MIM: 613620 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD2BP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD2BP2 - CD2 cytoplasmic tail binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D10B - TBC1 domain family member 10B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015527.3 | 1688 | Silent Mutation | ACC,ACG | T,T 601 | NP_056342.3 | |
XM_011545789.1 | 1688 | Silent Mutation | ACC,ACG | T,T 496 | XP_011544091.1 | |
XM_011545790.2 | 1688 | UTR 3 | XP_011544092.1 |