Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTGAAAGGCTGTCCACCTTGGTT[C/G]AATGTCAGCTTGTACCGTAAGCGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606005 | ||||||||||||||||||||
Literature Links: |
GGA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GGA2 - golgi associated, gamma adaptin ear containing, ARF binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015044.4 | 1850 | Missense Mutation | TTC,TTG | F,L 588 | NP_055859.1 | |
XM_017023074.1 | 1850 | Missense Mutation | TTC,TTG | F,L 551 | XP_016878563.1 | |
XM_017023075.1 | 1850 | Missense Mutation | TTC,TTG | F,L 501 | XP_016878564.1 |