Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610886 MIM: 605431 MIM: 611994 MIM: 601817 MIM: 610779 MIM: 611659 | ||||||||||||||||||||
Literature Links: |
EME2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EME2 - essential meiotic structure-specific endonuclease subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257370.1 | 911 | Intron | NP_001244299.1 | |||
XM_017023025.1 | 911 | Intron | XP_016878514.1 | |||
XM_017023026.1 | 911 | Intron | XP_016878515.1 |
MAPK8IP3 - mitogen-activated protein kinase 8 interacting protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPS34 - mitochondrial ribosomal protein S34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NME3 - NME/NM23 nucleoside diphosphate kinase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUBP2 - nucleotide binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPSB3 - splA/ryanodine receptor domain and SOCS box containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324081.1 | 911 | Missense Mutation | NP_001311010.1 | |||
NM_080861.3 | 911 | Missense Mutation | NP_543137.2 | |||
XM_005255673.4 | 911 | Missense Mutation | XP_005255730.1 |