Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACATGGAATTCCAGTAGATTAA[C/T]AGCACCCCCCGCCCACACACCTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616164 | ||||||||||||||||||||
Literature Links: |
SDR42E1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SDR42E1 - short chain dehydrogenase/reductase family 42E, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145168.2 | 9215 | Intron | NP_660151.2 | |||
XM_005256257.3 | 9215 | UTR 3 | XP_005256314.1 | |||
XM_011523471.2 | 9215 | UTR 3 | XP_011521773.1 | |||
XM_011523472.2 | 9215 | UTR 3 | XP_011521774.1 |