Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCATGCCCAGACATATGTCCCCC[A/G]GCTGGTAAGGATGAAGGTGTTCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613276 | ||||||||||||||||||||
Literature Links: |
USB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USB1 - U6 snRNA biogenesis phosphodiesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195302.1 | 412 | Missense Mutation | CAG,CGG | Q,R 112 | NP_001182231.1 | |
NM_001204911.1 | 412 | Missense Mutation | CAG,CGG | Q,R 112 | NP_001191840.1 | |
NM_024598.3 | 412 | Missense Mutation | CAG,CGG | Q,R 112 | NP_078874.2 | |
XM_005256144.3 | 412 | Missense Mutation | CAG,CGG | Q,R 61 | XP_005256201.1 | |
XM_011523328.2 | 412 | Missense Mutation | CAG,CGG | Q,R 99 | XP_011521630.1 | |
XM_011523329.2 | 412 | Missense Mutation | CAG,CGG | Q,R 61 | XP_011521631.1 | |
XM_011523330.1 | 412 | Missense Mutation | CAG,CGG | Q,R 112 | XP_011521632.1 |
ZNF319 - zinc finger protein 319 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |