Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTTTGATTTTTCCATGTTCTTT[A/G]CATCTAGATTGAAACACATCACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615624 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRNDE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRNDE - colorectal neoplasia differentially expressed (non-protein coding) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308963.1 | 1678 | Silent Mutation | TGC,TGT | C,C 61 | NP_001295892.1 | |
XM_017023562.1 | 1678 | Silent Mutation | TGC,TGT | C,C 61 | XP_016879051.1 | |
XM_017023563.1 | 1678 | Silent Mutation | TGC,TGT | C,C 61 | XP_016879052.1 |