Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCACCAAGTCGGGAGCCAGCTA[C/T]GCTTATATTCTAGAGGCCTTTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605641 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC7A6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC7A6 - solute carrier family 7 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076785.2 | 1119 | Silent Mutation | TAC,TAT | Y,Y 112 | NP_001070253.1 | |
NM_003983.5 | 1119 | Silent Mutation | TAC,TAT | Y,Y 112 | NP_003974.3 | |
XM_011523433.1 | 1119 | Silent Mutation | TAC,TAT | Y,Y 112 | XP_011521735.1 | |
XM_011523434.1 | 1119 | Silent Mutation | TAC,TAT | Y,Y 112 | XP_011521736.1 | |
XM_011523438.1 | 1119 | Silent Mutation | TAC,TAT | Y,Y 112 | XP_011521740.1 | |
XM_011523439.1 | 1119 | UTR 5 | XP_011521741.1 | |||
XM_017023851.1 | 1119 | Intron | XP_016879340.1 |