Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGGCGGCCATGGCTTCGGCTTCT[C/G]AGGGCGCTGACGACGACGGCAGTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604905 | ||||||||||||||||||||
Literature Links: |
ADAD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAD2 - adenosine deaminase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145400.1 | 130 | Missense Mutation | CAG,GAG | Q,E 6 | NP_001138872.1 | |
NM_139174.3 | 130 | Missense Mutation | CAG,GAG | Q,E 6 | NP_631913.3 | |
XM_005255814.1 | 130 | Missense Mutation | CAG,GAG | Q,E 6 | XP_005255871.1 | |
XM_011522919.1 | 130 | Missense Mutation | CAG,GAG | Q,E 6 | XP_011521221.1 | |
XM_011522920.1 | 130 | Intron | XP_011521222.1 |
TAF1C - TATA-box binding protein associated factor, RNA polymerase I subunit C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |