Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGGAGGGGCAGCTGTAGAGAAC[G/T]CGAAGCGGGAACACAGGGAGAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604567 MIM: 615175 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf92 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C16orf92 - chromosome 16 open reading frame 92 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109659.1 | Intron | NP_001103129.1 | ||||
NM_001109660.1 | Intron | NP_001103130.1 |
DOC2A - double C2 domain alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM57B - family with sequence similarity 57 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318504.1 | Intron | NP_001305433.1 | ||||
NM_031478.5 | Intron | NP_113666.2 | ||||
XM_005255613.3 | Intron | XP_005255670.1 | ||||
XM_005255614.3 | Intron | XP_005255671.1 | ||||
XM_005255615.2 | Intron | XP_005255672.1 | ||||
XM_017023749.1 | Intron | XP_016879238.1 | ||||
XM_017023750.1 | Intron | XP_016879239.1 | ||||
XM_017023751.1 | Intron | XP_016879240.1 | ||||
XM_017023752.1 | Intron | XP_016879241.1 | ||||
XM_017023753.1 | Intron | XP_016879242.1 | ||||
XM_017023754.1 | Intron | XP_016879243.1 |