Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTTAGTGAAGCCTCTCTCACAG[G/T]GAGAACACCTGTAAGGCTTTTCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZSCAN32 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZSCAN32 - zinc finger and SCAN domain containing 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284527.1 | 2257 | Missense Mutation | CAC,CCC | H,P 665 | NP_001271456.1 | |
NM_001284528.1 | 2257 | Missense Mutation | CAC,CCC | H,P 376 | NP_001271457.1 | |
NM_001284529.1 | 2257 | Missense Mutation | CAC,CCC | H,P 376 | NP_001271458.1 | |
NM_001324340.1 | 2257 | Missense Mutation | CAC,CCC | H,P 453 | NP_001311269.1 | |
NM_001324341.1 | 2257 | Missense Mutation | CAC,CCC | H,P 453 | NP_001311270.1 | |
NM_001324342.1 | 2257 | Missense Mutation | CAC,CCC | H,P 453 | NP_001311271.1 | |
NM_001324343.1 | 2257 | Missense Mutation | CAC,CCC | H,P 532 | NP_001311272.1 | |
NM_001324344.1 | 2257 | Missense Mutation | CAC,CCC | H,P 453 | NP_001311273.1 | |
NM_001324345.1 | 2257 | Missense Mutation | CAC,CCC | H,P 453 | NP_001311274.1 | |
NM_001324346.1 | 2257 | Missense Mutation | CAC,CCC | H,P 592 | NP_001311275.1 | |
NM_017810.3 | 2257 | Missense Mutation | CAC,CCC | H,P 453 | NP_060280.2 | |
XM_005255403.2 | 2257 | Missense Mutation | CAC,CCC | H,P 578 | XP_005255460.1 | |
XM_011522555.2 | 2257 | Missense Mutation | CAC,CCC | H,P 665 | XP_011520857.1 | |
XM_017023371.1 | 2257 | Missense Mutation | CAC,CCC | H,P 665 | XP_016878860.1 | |
XM_017023372.1 | 2257 | Missense Mutation | CAC,CCC | H,P 578 | XP_016878861.1 | |
XM_017023373.1 | 2257 | Missense Mutation | CAC,CCC | H,P 578 | XP_016878862.1 |