Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTTTGGCCAAAGAAGATTATTA[C/T]CAGATATTAGGAGTGCCTCGAAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608382 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJA3 - DnaJ heat shock protein family (Hsp40) member A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135110.2 | 362 | Silent Mutation | TAC,TAT | Y,Y 95 | NP_001128582.1 | |
NM_001286516.1 | 362 | Intron | NP_001273445.1 | |||
NM_005147.5 | 362 | Silent Mutation | TAC,TAT | Y,Y 95 | NP_005138.3 |