Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCGCGCTGGTGGTGCACCCGGG[C/T]ACGGCACGGCTGGGGAGCCCGGACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186591 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928762 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101928762 - uncharacterized LOC101928762 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STX4 - syntaxin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272095.1 | 405 | UTR 5 | NP_001259024.1 | |||
NM_001272096.1 | 405 | Missense Mutation | GCA,GTA | A,V 21 | NP_001259025.1 | |
NM_004604.4 | 405 | Silent Mutation | GGC,GGT | G,G 30 | NP_004595.2 | |
XM_005255521.1 | 405 | Missense Mutation | GCA,GTA | A,V 19 | XP_005255578.1 | |
XM_011545925.2 | 405 | Intron | XP_011544227.1 |