Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCAGGCACGTCTTTGCTGGCTGA[A/G]TCTGAGTGCATTTCAGGAAAGATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608293 MIM: 610238 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP17 - Rho GTPase activating protein 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006634.2 | 2595 | Silent Mutation | GAC,GAT | D,D 858 | NP_001006635.1 | |
NM_018054.5 | 2595 | Silent Mutation | GAC,GAT | D,D 780 | NP_060524.4 | |
XM_011545873.2 | 2595 | Silent Mutation | GAC,GAT | D,D 890 | XP_011544175.1 | |
XM_011545874.2 | 2595 | Silent Mutation | GAC,GAT | D,D 889 | XP_011544176.1 | |
XM_011545875.2 | 2595 | UTR 3 | XP_011544177.1 | |||
XM_011545876.2 | 2595 | UTR 3 | XP_011544178.1 | |||
XM_011545877.2 | 2595 | Silent Mutation | GAC,GAT | D,D 813 | XP_011544179.1 | |
XM_011545878.2 | 2595 | Silent Mutation | GAC,GAT | D,D 812 | XP_011544180.1 | |
XM_017023390.1 | 2595 | UTR 3 | XP_016878879.1 |
SLC5A11 - solute carrier family 5 member 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |