Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTTACACCAGAAAGAACCCTAC[A/C]ATCCACAGAGGGCCGCACAGAGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615153 MIM: 614151 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLKL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MLKL - mixed lineage kinase domain-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142497.2 | 2079 | Nonsense Mutation | GAA,TAA | E,* 253 | NP_001135969.1 | |
NM_152649.3 | 2079 | Nonsense Mutation | GAA,TAA | E,* 461 | NP_689862.1 | |
XM_005255834.3 | 2079 | Nonsense Mutation | GAA,TAA | E,* 461 | XP_005255891.1 | |
XM_011522936.2 | 2079 | Intron | XP_011521238.1 |
RFWD3 - ring finger and WD repeat domain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |