Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGACCAGGGCAGCACTCCTCCT[A/G]TTCACAGGTGAGCCTGGACCCCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151510 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGAX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITGAX - integrin subunit alpha X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000887.4 | 151 | Silent Mutation | CTA,CTG | L,L 10 | NP_000878.2 | |
NM_001286375.1 | 151 | Silent Mutation | CTA,CTG | L,L 10 | NP_001273304.1 | |
XM_011545852.1 | 151 | Silent Mutation | CTA,CTG | L,L 10 | XP_011544154.1 | |
XM_011545854.1 | 151 | Silent Mutation | CTA,CTG | L,L 10 | XP_011544156.1 |