Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATGGGTGGCAAGTCTAATCTAT[C/T]CGAATCTTTACAGGCACTTTTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF688 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF688 - zinc finger protein 688 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024683.1 | 1090 | UTR 3 | NP_001019854.1 | |||
NM_145271.3 | 1090 | UTR 3 | NP_660314.1 | |||
XM_005255139.2 | 1090 | UTR 3 | XP_005255196.1 | |||
XM_005255140.2 | 1090 | UTR 3 | XP_005255197.1 |
ZNF785 - zinc finger protein 785 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |